Learn More
Strem, An Ascensus Company CAS# 19132-06-0. 100mg. (2S,3S)-(+)-2,3-Butanediol, min. 97%. MFCD00063648
SDP
Catalog No. 5090109168
Encompass
CAS# 19132-06-0. 100mg. (2S,3S)-(+)-2,3-Butanediol, min. 97%. MFCD00063648. Molecular Weight: 90.12. Molecular Formula: CH3CH(OH)CH(OH)CH3. Color/form: colorless liq. Strem# 08-0201. http://www.strem.com/catalog/v/08-0201/
Catalog No. 50-901-09168 Supplier Strem, An Ascensus Company Supplier No. 080201100MG
CAS# 19132-06-0. 100mg. (2S,3S)-(+)-2,3-Butanediol, min. 97%. MFCD00063648. Molecular Weight: 90.12. Molecular Formula: CH3CH(OH)CH(OH)CH3. Color/form: colorless liq. Strem# 08-0201. http://www.strem.com/catalog/v/08-0201/