Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2S,3S,5R,6R)-5,6-Bis(hydroxymethyl)-2,3-dimethoxy-2,3-dimethyl-1,4-dioxane 98.0+%, TCI America™
SDP

Catalog No. B3696200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B3696200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B3696200MG Supplier TCI America Supplier No. B3696200MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 241811-66-5
Molecular Formula C10H20O6
Molecular Weight (g/mol) 236.264
InChI Key RAIJJZVROJTPEU-IMSYWVGJSA-N
Synonym (2R,3R,5S,6S)-5,6-Dimethoxy-5,6-dimethyl-1,4-dioxane-2,3-dimethanol
PubChem CID 10585874
IUPAC Name [(2R,3R,5S,6S)-3-(hydroxymethyl)-5,6-dimethoxy-5,6-dimethyl-1,4-dioxan-2-yl]methanol
SMILES CC1(C(OC(C(O1)CO)CO)(C)OC)OC

Specifications

Melting Point 120°C
Color White-Yellow
Quantity 200 mg
Physical Form Crystal-Powder at 20°C
Formula Weight 236.26
Percent Purity ≥98.0% (GC)
Chemical Name or Material (2S,3S,5R,6R)-5,6-Bis(hydroxymethyl)-2,3-dimethoxy-2,3-dimethyl-1,4-dioxane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.