Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (2S,3S)-(-)-Bis(Diphenylphosphino)Butane

Catalog No. 2590981G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-909-81G 1 g
25-909-8250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 25-909-81G Supplier Sigma Aldrich Supplier No. 2590981G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(2S,3S)-(-)-Bis(Diphenylphosphino)Butane

Specifications

CAS 64896-28-2
Melting Point 108°C to 110°C
Molecular Formula C28H28P2
Linear Formula [-CH(CH3)P(C6H5)2]2
MDL Number MFCD00009836
Quantity 1 g
Synonym (2S,3S)-(-)-2,3-Bis(diphenylphosphino)butane; (S,S)-Chiraphos
Molecular Weight (g/mol) 426.47 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.