Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2S,3S)-(-)-Bis(diphenylphosphino)butane, 98%

Catalog No. AAH27586MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH27586MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH27586MD Supplier Thermo Scientific Chemicals Supplier No. H27586MD

CAS: 64896-28-2 | C28H28P2 | 426.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64896-28-2
Molecular Formula C28H28P2
Molecular Weight (g/mol) 426.48
MDL Number MFCD00009836
InChI Key FWXAUDSWDBGCMN-ZBXWWWQKNA-N
Synonym s,s-chiraphos, 2s,3s---bis diphenylphosphino butane, chiraphos, chiraphos, s,s, unii-6qr78gzl9b, 6qr78gzl9b, 2s,3s-butane-2,3-diylbis diphenylphosphine, phosphine, 1s,2s-1,2-dimethyl-1,2-ethanediyl bis diphenyl, --chiraphos, 2s,3s-chiraphos
PubChem CID 10113249
IUPAC Name [(2S,3S)-3-diphenylphosphanylbutan-2-yl]-diphenylphosphane
SMILES C[C@@H]([C@H](C)P(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 107°C to 109°C
Quantity 250 mg
Beilstein 4268869
Sensitivity Air Sensitive
Formula Weight 426.48
Percent Purity 98%
Chemical Name or Material (2S,3S)-(-)-Bis(diphenylphosphino)butane
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.