Learn More
TARGETMOL CHEMICALS INC 2S-4-BIPHENYL-4-YL-N-HYD 5MG
SDP
Catalog No. 502579638
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (2S)-4-biphenyl-4-yl-N-hydroxy-2-methyl-2-(methylsulfonyl)butanamide is a LpxC inhibitor with IC50 >95.2 nM. purity: 99%
Catalog No. 50-257-9638 Supplier TARGETMOL CHEMICALS INC Supplier No. T602055MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (2S)-4-biphenyl-4-yl-N-hydroxy-2-methyl-2-(methylsulfonyl)butanamide is a LpxC inhibitor with IC50 >95.2 nM. purity: 99%