Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (2S,4R)-1-[(2S)-2-[[2-[2-[2-[2-(2-aminoethoxy)ethoxy]ethoxy]ethoxy]acetyl]amino]-3,3-dimethyl... | 2010159-57-4 | MFCD31657338 | 99.5% | 663.83 | C32H49N5O8S | 5 MG
SDP

Catalog No. 5000244985
Encompass_Preferred

(S,R,S)-AHPC-PEG4-NH2 is a VHL E3 ligase ligand-linker conjugate that combines an (S,R,S)-AHPC VHL ligand with a four-unit polyethylene glycol (PEG4) linker. It is supplied as a white to off-white solid and is used as a building block in proteolysis targeting chimera (PROTAC) design to recruit the VHL E3 ligase.

  • Used as a VHL ligand-linker component in PROTAC synthesis and E3 ligase recruitment studies.
  • Purity (LCMS) 99.51%.
  • Molecular weight 663.83 (free base).
  • Chemical formula C32H49N5O8S.
  • Form white to off-white solid.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (2 years) or -20°C (1 year).

Catalog No. 50-002-44985 Supplier Medchemexpress LLC Supplier No. HY103604A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S,R,S)-AHPC-PEG4-NH2 is a VHL E3 ligase ligand-linker conjugate that combines an (S,R,S)-AHPC VHL ligand with a four-unit polyethylene glycol (PEG4) linker. It is supplied as a white to off-white solid and is used as a building block in proteolysis targeting chimera (PROTAC) design to recruit the VHL E3 ligase.

  • Used as a VHL ligand-linker component in PROTAC synthesis and E3 ligase recruitment studies.
  • Purity (LCMS) 99.51%.
  • Molecular weight 663.83 (free base).
  • Chemical formula C32H49N5O8S.
  • Form white to off-white solid.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (2 years) or -20°C (1 year).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.