Learn More
Medchemexpress LLC (2S,4R)-1-((9H-Fluoren-9-yl)methyl) 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylate | 122350-59-8 | C21H21NO5 | 10 G
SDP
Catalog No. 5000434418
Encompass_Preferred
(2S,4R)-1-((9H-Fluoren-9-yl)methyl) 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylate is a proline derivative recognized as a beneficial ergogenic dietary substance. It is used as an ergogenic supplement, influencing anabolic hormone secretion, supplying fuel during exercise, enhancing mental performance, and preventing exercise-induced muscle damage.
  • Influences anabolic hormone secretion
  • Supplies fuel during exercise
  • Enhances mental performance during stress-related tasks
  • Prevents exercise-induced muscle damage
  • Stable as powder at -20°C for 3 years and 4°C for 2 years
  • Stable in solvent at -80°C for 6 months and -20°C for 1 month
Catalog No. 50-004-34418 Supplier Medchemexpress LLC Supplier No. HY7912310G
(2S,4R)-1-((9H-Fluoren-9-yl)methyl) 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylate is a proline derivative recognized as a beneficial ergogenic dietary substance. It is used as an ergogenic supplement, influencing anabolic hormone secretion, supplying fuel during exercise, enhancing mental performance, and preventing exercise-induced muscle damage.
  • Influences anabolic hormone secretion
  • Supplies fuel during exercise
  • Enhances mental performance during stress-related tasks
  • Prevents exercise-induced muscle damage
  • Stable as powder at -20°C for 3 years and 4°C for 2 years
  • Stable in solvent at -80°C for 6 months and -20°C for 1 month