Learn More
Medchemexpress LLC (2S,4R)-1-((9H-Fluoren-9-yl)methyl) 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylate | 122350-59-8 | C21H21NO5 | 100 G
SDP
Catalog No. 5000433024
Encompass_Preferred
(2S,4R)-1-((9H-Fluoren-9-yl)methyl) 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylate is a proline derivative with commercial applications as an ergogenic supplement. It influences anabolic hormone secretion, supplies fuel during exercise, impacts mental performance during stressful tasks, and prevents exercise-induced muscle damage.
  • Proline derivative
  • Influences anabolic hormone secretion
  • Supplies fuel during exercise
  • Impacts mental performance during stressful tasks
  • Prevents exercise-induced muscle damage
  • Store as powder at -20°C for up to 3 years or 4°C for up to 2 years
  • Store in solvent at -80°C for up to 6 months or -20°C for up to 1 month
  • Soluble in DMSO at 100 mg/mL (272.18 mM)
Catalog No. 50-004-33024 Supplier Medchemexpress LLC Supplier No. HY79123100G
(2S,4R)-1-((9H-Fluoren-9-yl)methyl) 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylate is a proline derivative with commercial applications as an ergogenic supplement. It influences anabolic hormone secretion, supplies fuel during exercise, impacts mental performance during stressful tasks, and prevents exercise-induced muscle damage.
  • Proline derivative
  • Influences anabolic hormone secretion
  • Supplies fuel during exercise
  • Impacts mental performance during stressful tasks
  • Prevents exercise-induced muscle damage
  • Store as powder at -20°C for up to 3 years or 4°C for up to 2 years
  • Store in solvent at -80°C for up to 6 months or -20°C for up to 1 month
  • Soluble in DMSO at 100 mg/mL (272.18 mM)