Learn More
Medchemexpress LLC (2S,4R)-1-((S)-14-Azido-2-(tert-butyl)-4-oxo-6,9,12-trioxa-3-azatetradecan-1-oyl)-4-hydroxy-N-(4-( | 1797406-80-4 | MFCD31807299 | >95.0% | 645.77 g/mol | C30H43N7O7S | 5 MG
SDP
Catalog No. 5000245012
Encompass_Preferred
(S,R,S)-AHPC-PEG3-N3 is an azide-terminated VHL ligand-linker conjugate that combines an (S,R,S)-AHPC VHL ligand with a three-unit polyethylene glycol linker. It is intended for use in PROTAC design, click-chemistry conjugation, and other chemical biology applications, and is supplied in small milligram quantities with manufacturer-verified purity and storage recommendations.
  • Provides an azide-terminated PEG3 linker for click chemistry conjugation.
  • Designed to recruit the von Hippel-Lindau (VHL) E3 ligase for targeted protein degradation studies.
  • Supplied in accurately weighed milligram quantities suitable for medicinal chemistry workflows.
  • Manufacturer certificate of analysis confirms purity and storage conditions for long-term stability.
Catalog No. 50-002-45012 Supplier Medchemexpress LLC Supplier No. HY1035985MG
(S,R,S)-AHPC-PEG3-N3 is an azide-terminated VHL ligand-linker conjugate that combines an (S,R,S)-AHPC VHL ligand with a three-unit polyethylene glycol linker. It is intended for use in PROTAC design, click-chemistry conjugation, and other chemical biology applications, and is supplied in small milligram quantities with manufacturer-verified purity and storage recommendations.
  • Provides an azide-terminated PEG3 linker for click chemistry conjugation.
  • Designed to recruit the von Hippel-Lindau (VHL) E3 ligase for targeted protein degradation studies.
  • Supplied in accurately weighed milligram quantities suitable for medicinal chemistry workflows.
  • Manufacturer certificate of analysis confirms purity and storage conditions for long-term stability.