Learn More
Medchemexpress LLC (2S,4R)-1-((S)-2-(1-cyanocyclopropane-1-carboxamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylt | 2097381-85-4 | MFCD30742947 | 99.5% | 523.65 g/mol | C27H33N5O4S | 50 MG
SDP
Catalog No. 5000213012
Encompass_Preferred
VH-298 is a cell-permeable inhibitor of the VHL:HIF-α interaction that induces HIF-α accumulation and serves as an E3 ligase ligand for PROTAC synthesis. It is potent (Kd 80-90 nM), shows minimal off-target activity in broad panels, and is suitable as a chemical probe for studying hypoxic response and targeted protein degradation.
  • Highly potent VHL:HIF-α interaction inhibitor (Kd 80-90 nM).
  • Induces HIF-α accumulation in human cell lines.
  • Functions as an E3 ligase ligand for PROTAC synthesis.
  • Cell-permeable with low cellular toxicity.
  • Minimal off-target activity across kinase, GPCR, and ion channel panels at 50 μM.
  • High purity and defined storage and solubility profiles for experimental use.
Catalog No. 50-002-13012 Supplier Medchemexpress LLC Supplier No. HY10094750MG
VH-298 is a cell-permeable inhibitor of the VHL:HIF-α interaction that induces HIF-α accumulation and serves as an E3 ligase ligand for PROTAC synthesis. It is potent (Kd 80-90 nM), shows minimal off-target activity in broad panels, and is suitable as a chemical probe for studying hypoxic response and targeted protein degradation.
  • Highly potent VHL:HIF-α interaction inhibitor (Kd 80-90 nM).
  • Induces HIF-α accumulation in human cell lines.
  • Functions as an E3 ligase ligand for PROTAC synthesis.
  • Cell-permeable with low cellular toxicity.
  • Minimal off-target activity across kinase, GPCR, and ion channel panels at 50 μM.
  • High purity and defined storage and solubility profiles for experimental use.