Learn More
Medchemexpress LLC (2S,4R)-1-((S)-2-(tert-butyl)-14-(4-(4-((3-chloro-4-((3-fluorobenzyl)oxy)phenyl)amino)quinazolin-6-yl) | 2230821-40-4 | 99.7% | 1074.65 | C57H61ClFN7O9S | 5 MG
SDP
Catalog No. 5000262817
Encompass_Preferred
SJF-1521 is a small-molecule PROTAC degrader that selectively targets epidermal growth factor receptor (EGFR) for proteasomal degradation. It is intended for research applications investigating EGFR signaling, degradation mechanisms, and tumor biology, and is supplied with manufacturer analytical data supporting its characterization.
  • Selective EGFR-targeting PROTAC degrader.
  • Demonstrated to induce EGFR degradation in OVCAR8 cells.
  • High reported purity (≈99.7%).
  • Characterized by molecular weight and structural analytical data.
  • Supplied in small research quantities suitable for in vitro studies.
Catalog No. 50-002-62817 Supplier Medchemexpress LLC Supplier No. HY1318655MG
Add to Cart
Add to Cart
SJF-1521 is a small-molecule PROTAC degrader that selectively targets epidermal growth factor receptor (EGFR) for proteasomal degradation. It is intended for research applications investigating EGFR signaling, degradation mechanisms, and tumor biology, and is supplied with manufacturer analytical data supporting its characterization.
  • Selective EGFR-targeting PROTAC degrader.
  • Demonstrated to induce EGFR degradation in OVCAR8 cells.
  • High reported purity (≈99.7%).
  • Characterized by molecular weight and structural analytical data.
  • Supplied in small research quantities suitable for in vitro studies.