Learn More
Medchemexpress LLC (2S,4R)-4-hydroxypyrrolidine-2-carboxylic-2,4,5,5-d4 acid | 2734920-51-3 | 99.9% | 135.15 | C5H5D4NO3 | 1 MG
SDP
Catalog No. 5000284033
Encompass_Preferred
Deuterium-labeled L-hydroxyproline used as a chiral building block and as an analytical internal standard for quantitative methods. Supplied as a white to off-white solid with high isotopic enrichment and high chemical purity, the compound is intended for research and analytical applications where labeled internal standards are required.
  • Deuterium labeled at positions 2, 4, 5, and 5.
  • High purity (99.9% HPLC) and high isotopic enrichment.
  • Suitable as tracer and internal standard for NMR, GC-MS, and LC-MS quantitative analysis.
  • White to off-white solid; stable when stored as powder at -20 °C or in solution at -80 °C.
  • Available in small mg quantities (1 MG, 5 MG, 10 MG) for analytical use.
Catalog No. 50-002-84033 Supplier Medchemexpress LLC Supplier No. HYW6542321MG
Deuterium-labeled L-hydroxyproline used as a chiral building block and as an analytical internal standard for quantitative methods. Supplied as a white to off-white solid with high isotopic enrichment and high chemical purity, the compound is intended for research and analytical applications where labeled internal standards are required.
  • Deuterium labeled at positions 2, 4, 5, and 5.
  • High purity (99.9% HPLC) and high isotopic enrichment.
  • Suitable as tracer and internal standard for NMR, GC-MS, and LC-MS quantitative analysis.
  • White to off-white solid; stable when stored as powder at -20 °C or in solution at -80 °C.
  • Available in small mg quantities (1 MG, 5 MG, 10 MG) for analytical use.