Learn More
Medchemexpress LLC (2S,4R)-4-methylglutamic acid | 31137-74-3 | MFCD00937765 | ≥97.0% | 161.16 g/mol | C6H11NO4 | 100 MG
SDP
Catalog No. 5000212658
Encompass_Preferred
SYM 2081 ((2S,4R)-4-methylglutamic acid, CAS 31137-74-3) is a selective kainate receptor agonist used as a research tool in neuroscience and pharmacology. It modulates glutamate transport and receptor activity, and has been employed in studies of neuroprotection and inflammatory or neuropathic pain. Supplied as a solid for laboratory use, it is typically stored at -20°C.
  • Selective kainate receptor agonist for receptor pharmacology studies.
  • Modulates glutamate transport and excitatory amino acid uptake.
  • Applied in models of neuroprotection and pain research.
  • Suitable for in vitro and in vivo experimental use in small research quantities.
  • Solid form with recommended storage at -20°C to preserve stability.
Catalog No. 50-002-12658 Supplier Medchemexpress LLC Supplier No. HY101310100MG
SYM 2081 ((2S,4R)-4-methylglutamic acid, CAS 31137-74-3) is a selective kainate receptor agonist used as a research tool in neuroscience and pharmacology. It modulates glutamate transport and receptor activity, and has been employed in studies of neuroprotection and inflammatory or neuropathic pain. Supplied as a solid for laboratory use, it is typically stored at -20°C.
  • Selective kainate receptor agonist for receptor pharmacology studies.
  • Modulates glutamate transport and excitatory amino acid uptake.
  • Applied in models of neuroprotection and pain research.
  • Suitable for in vitro and in vivo experimental use in small research quantities.
  • Solid form with recommended storage at -20°C to preserve stability.