Learn More
Enzo Life Sciences (2S,4R)-4-Methylglutamic acid (5mg). CAS: 31137-74-3
SDP
Supplier:  Enzo Life Sciences ALX550186M005
Encompass_Preferred
Potent and highly selective kainate receptor agonist, with an IC50 for inhibition of [3H]-kainate binding of 35 nM and almost 3,000- and 200-fold selectivity for kainate receptors over AMPA and NMDA receptors respectively. Also selectively inhibits the cloned excitatory amino acid transporter EAAT2 at higher concentrations. Alternative name: SYM 2081. Purity: ≥97%. Solubility: Soluble in dilute aqueous acid. Long Term Storage: -20°C.
Catalog No. 50-200-9496