Learn More
Medchemexpress LLC (2S,4S)-1-((S)-2-amino-3,3-dimethylbutanoyl)-4-hydroxy-N-((S)-1-(4-(4-methylthiazol-5-yl)phenyl) | 2380273-26-5 | 98.6% | 481.05 | C23H33ClN4O3S | 5 G
SDP
Catalog No. 5000284326
Encompass_Preferred
(2S,4S)-1-((S)-2-Amino-3,3-dimethylbutanoyl)-4-hydroxy-N-((S)-1-(4-(4-methylthiazol-5-yl)phenyl)ethyl)pyrrolidine-2-carboxamide hydrochloride is a chiral pyrrolidine-derived carboxamide supplied as the hydrochloride salt for laboratory research and synthetic chemistry. The compound is provided as a solid, with molecular formula C23H33ClN4O3S, a molecular weight of 481.05 g/mol, and is characterized by manufacturer documentation including COA and SDS.
  • Chiral pyrrolidine carboxamide suitable for synthesis and analytical studies.
  • Provided as hydrochloride salt in solid form.
  • Characterized by COA with reported purity 98.6% (LCMS).
  • Recommended storage: -20°C, sealed, protect from moisture and light.
  • Intended for laboratory research use only; not for human use.
Catalog No. 50-002-84326 Supplier Medchemexpress LLC Supplier No. HYW6743505G
(2S,4S)-1-((S)-2-Amino-3,3-dimethylbutanoyl)-4-hydroxy-N-((S)-1-(4-(4-methylthiazol-5-yl)phenyl)ethyl)pyrrolidine-2-carboxamide hydrochloride is a chiral pyrrolidine-derived carboxamide supplied as the hydrochloride salt for laboratory research and synthetic chemistry. The compound is provided as a solid, with molecular formula C23H33ClN4O3S, a molecular weight of 481.05 g/mol, and is characterized by manufacturer documentation including COA and SDS.
  • Chiral pyrrolidine carboxamide suitable for synthesis and analytical studies.
  • Provided as hydrochloride salt in solid form.
  • Characterized by COA with reported purity 98.6% (LCMS).
  • Recommended storage: -20°C, sealed, protect from moisture and light.
  • Intended for laboratory research use only; not for human use.