Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (2S,4S)-1-((S)-2-amino-3,3-dimethylbutanoyl)-4-hydroxy-N-((S)-1-(4-(4-methylthiazol-5-yl)phenyl)ethyl | 2380273-26-5 | MFCD32876834 | 98.6% | 481.05 g/mol | C23H33ClN4O3S | 500 MG
SDP

Supplier:  Medchemexpress LLC HYW674350500MG

Encompass_Preferred

(2S,4S)-1-((S)-2-amino-3,3-dimethylbutanoyl)-4-hydroxy-N-((S)-1-(4-(4-methylthiazol-5-yl)phenyl)ethyl)pyrrolidine-2-carboxamide hydrochloride is a research-grade small molecule supplied as the hydrochloride salt for biochemical and medicinal chemistry studies. It is characterized by a molecular formula C23H33ClN4O3S, molecular weight 481.05 g/mol, and LCMS purity 98.59%. Documentation available includes a certificate of analysis and a safety data sheet.

  • Hydrochloride salt suitable for research use.
  • Purity 98.6% (LCMS).
  • Molecular weight 481.05 g/mol, formula C23H33ClN4O3S.
  • Supplied with COA and SDS documentation.
  • Recommended storage: -20°C, sealed, protected from moisture and light.

Catalog No. 50-002-84043


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.