Learn More
Medchemexpress LLC (2S,4S)-1-((S)-2-amino-3,3-dimethylbutanoyl)-4-hydroxy-N-((S)-1-(4-(4-methylthiazol-5-yl)phenyl)ethyl)pyr | 2380273-26-5 | 98.6% | 481.05 g·mol⁻¹ | C23H33ClN4O3S | 100 MG

Supplier: Medchemexpress LLC HYW674350100MG
(2S,4S)-1-((S)-2-amino-3,3-dimethylbutanoyl)-4-hydroxy-N-((S)-1-(4-(4-methylthiazol-5-yl)phenyl)ethyl)pyrrolidine-2-carboxamide (hydrochloride) is a synthetic peptidomimetic research compound provided as a hydrochloride salt for laboratory research and analytical applications. The material is supplied as a solid with high reported purity; consult the SDS and COA for full handling and safety information.
- Purity 98.59%.
- Physical form solid.
- Salt form hydrochloride.
- Molecular weight 481.05 g·mol⁻¹.
- CAS number 2380273-26-5.
- Recommended storage -20°C, sealed and protected from moisture and light.
- Supplied in small research sizes, including 100 mg, 250 mg, 500 mg, and 1 g.
- Intended for research and analytical use only; follow SDS precautions.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.