Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (2S,4S)-1-((S)-2-amino-3,3-dimethylbutanoyl)-4-hydroxy pyrrolidine carboxamide hydrochloride | 2380273-26-5 | 98.6% | 481.05 g/mol | C23H33ClN4O3S | 250 MG
SDP

Catalog No. 5000284547
Encompass_Preferred

(2S,4S)-1-((S)-2-amino-3,3-dimethylbutanoyl)-4-hydroxy-N-((S)-1-(4-(4-methylthiazol-5-yl)phenyl)ethyl)pyrrolidine-2-carboxamide hydrochloride is a synthetic peptidomimetic intermediate used in pharmaceutical research and synthesis. It is supplied as a white to yellow solid with molecular weight 481.05 g/mol and CAS 2380273-26-5; batch COA reports LCMS purity 98.59%. Recommended storage is -20°C, sealed, protected from moisture and light.

  • Hydrochloride salt suitable for synthetic intermediates
  • High purity (98.6%) by LCMS
  • White to yellow solid appearance
  • Molecular weight 481.05 g/mol; CAS 2380273-26-5
  • Intended for use as a drug intermediate and research chemical
  • Store at -20°C, sealed, protect from moisture and light

Catalog No. 50-002-84547 Supplier Medchemexpress LLC Supplier No. HYW674350250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(2S,4S)-1-((S)-2-amino-3,3-dimethylbutanoyl)-4-hydroxy-N-((S)-1-(4-(4-methylthiazol-5-yl)phenyl)ethyl)pyrrolidine-2-carboxamide hydrochloride is a synthetic peptidomimetic intermediate used in pharmaceutical research and synthesis. It is supplied as a white to yellow solid with molecular weight 481.05 g/mol and CAS 2380273-26-5; batch COA reports LCMS purity 98.59%. Recommended storage is -20°C, sealed, protected from moisture and light.

  • Hydrochloride salt suitable for synthetic intermediates
  • High purity (98.6%) by LCMS
  • White to yellow solid appearance
  • Molecular weight 481.05 g/mol; CAS 2380273-26-5
  • Intended for use as a drug intermediate and research chemical
  • Store at -20°C, sealed, protect from moisture and light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.