Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (2S,4S)-1-tert-butyl 2-methyl-4-hydroxypiperidine-1,2-dicarboxylate | 254882-14-9 | 99.7% | 259.30 | 1 G
SDP

Catalog No. 5000402619
Encompass_Preferred

AN-12-H5 intermediate-1 ((2S,4S)-1-tert-butyl 2-methyl-4-hydroxypiperidine-1,2-dicarboxylate) is an intermediate of the viral inhibitor AN-12-H5 and can be used to synthesize Antibody-Drug Conjugates (ADCs).

  • Intermediate for viral inhibitor AN-12-H5.
  • Used to synthesize Antibody-Drug Conjugates (ADCs).
  • Targets replication processes of PV and enterovirus 71 (EV71).
  • Shows no inhibitory effect on PI4KB activity.
  • Moderate inhibitory effects on PI 3-kinase activity.

Catalog No. 50-004-02619 Supplier Medchemexpress LLC Supplier No. HY22055B1G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AN-12-H5 intermediate-1 ((2S,4S)-1-tert-butyl 2-methyl-4-hydroxypiperidine-1,2-dicarboxylate) is an intermediate of the viral inhibitor AN-12-H5 and can be used to synthesize Antibody-Drug Conjugates (ADCs).

  • Intermediate for viral inhibitor AN-12-H5.
  • Used to synthesize Antibody-Drug Conjugates (ADCs).
  • Targets replication processes of PV and enterovirus 71 (EV71).
  • Shows no inhibitory effect on PI4KB activity.
  • Moderate inhibitory effects on PI 3-kinase activity.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.