Learn More
Medchemexpress LLC (2S,4S)-5-([1,1'-Biphenyl]-4-yl)-4-((tert-butoxycarbonyl)amino)-2-methylpentanoic acid | 1012341-52-4 | 99.0% | C23H29NO4 | 50 MG
SDP
Catalog No. 5000434890
Encompass_Preferred
(2S,4S)-5-([1,1'-Biphenyl]-4-yl)-4-((tert-butoxycarbonyl)amino)-2-methylpentanoic acid is a drug intermediate for the synthesis of various active compounds. It is intended for research use only and not for sale to patients.
  • Drug intermediate for synthesis of various active compounds
  • For research use only
  • Appears as a solid
  • Storage as powder: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month
Catalog No. 50-004-34890 Supplier Medchemexpress LLC Supplier No. HY7883950MG
(2S,4S)-5-([1,1'-Biphenyl]-4-yl)-4-((tert-butoxycarbonyl)amino)-2-methylpentanoic acid is a drug intermediate for the synthesis of various active compounds. It is intended for research use only and not for sale to patients.
  • Drug intermediate for synthesis of various active compounds
  • For research use only
  • Appears as a solid
  • Storage as powder: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month