Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (2S,4S)-pyrrolidine-2-carboxamide derivative (diTFA) | 00-00-0 | 98.1% | 1279.84 g·mol⁻¹ | C61H78ClF7N10O8S | 5 MG
SDP

Catalog No. 5000221240
Encompass_Preferred

(4S)-PROTAC SOS1 degrader-1 (diTFA) is a research-grade PROTAC designed to induce targeted degradation of the SOS1 protein in cellular studies. It reduces SOS1 levels and downstream signaling pathways such as pERK and RAS-GTP, and is supplied as a TFA salt for preclinical research applications.

  • Potent SOS1 degrader in cell-based assays.
  • Reduces pERK and RAS-GTP signaling.
  • High purity (~98.1%).
  • Molecular weight 1279.84 g·mol⁻¹.
  • Supplied as a 5 mg vial for research use.
  • Certificate of analysis and safety data sheet available.
  • Store at -20 °C, sealed, to preserve stability.

Catalog No. 50-002-21240 Supplier Medchemexpress LLC Supplier No. HY144657A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(4S)-PROTAC SOS1 degrader-1 (diTFA) is a research-grade PROTAC designed to induce targeted degradation of the SOS1 protein in cellular studies. It reduces SOS1 levels and downstream signaling pathways such as pERK and RAS-GTP, and is supplied as a TFA salt for preclinical research applications.

  • Potent SOS1 degrader in cell-based assays.
  • Reduces pERK and RAS-GTP signaling.
  • High purity (~98.1%).
  • Molecular weight 1279.84 g·mol⁻¹.
  • Supplied as a 5 mg vial for research use.
  • Certificate of analysis and safety data sheet available.
  • Store at -20 °C, sealed, to preserve stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.