Learn More
Medchemexpress LLC (2S,4S)-Sacubitril | 149709-63-7 | C24H29NO5 | 1 ML
SDP
Catalog No. 5000435055
Encompass_Preferred
(2S,4S)-Sacubitril is an impurity of Sacubitril, a potent NEP inhibitor used for heart failure research. This compound is also a stereoisomer derived from Sacubitril.
  • Impurity of sacubitril
  • Stereoisomer derived from sacubitril
  • Appears as colorless to light yellow oil
  • Molecular weight: 411.49
Catalog No. 50-004-35055 Supplier Medchemexpress LLC Supplier No. HY7884110MM/1ML
(2S,4S)-Sacubitril is an impurity of Sacubitril, a potent NEP inhibitor used for heart failure research. This compound is also a stereoisomer derived from Sacubitril.
  • Impurity of sacubitril
  • Stereoisomer derived from sacubitril
  • Appears as colorless to light yellow oil
  • Molecular weight: 411.49