Learn More
Medchemexpress LLC (2S,5R)-6-(benzyloxy)-7-oxo-1,6-diazabicyclo[3.2.1]octane-2-carboxamide | 1192651-49-2 | MFCD30541561 | C14H17N3O3 | 100 MG
SDP
Catalog No. 5000433282
Encompass_Preferred
(2S,5R)-6-(benzyloxy)-7-oxo-1,6-diazabicyclo[3.2.1]octane-2-carboxamide is an impurity of Avibactam, a novel β-lactamase inhibitor with a non-lactam structural scaffold.
  • Avibactam irreversibly inhibits lactamase from Mycobacterium tuberculosis.
  • Synthesized using (2S,5R)-6-(benzyloxy)-7-oxo-1,6-diazabicyclo[3.2.1]octane-2-carboxylic acid as a raw material.
Catalog No. 50-004-33282 Supplier Medchemexpress LLC Supplier No. HY79858100MG
(2S,5R)-6-(benzyloxy)-7-oxo-1,6-diazabicyclo[3.2.1]octane-2-carboxamide is an impurity of Avibactam, a novel β-lactamase inhibitor with a non-lactam structural scaffold.
  • Avibactam irreversibly inhibits lactamase from Mycobacterium tuberculosis.
  • Synthesized using (2S,5R)-6-(benzyloxy)-7-oxo-1,6-diazabicyclo[3.2.1]octane-2-carboxylic acid as a raw material.