Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (2S,5S)-(-)-5-Benzyl-3-Methyl-2-(5-Methyl-2-Furyl)-4-Imidazolidinone

Catalog No. 668540250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
66-854-0250MG 250 mg
66-854-01G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 66-854-0250MG Supplier Sigma Aldrich Supplier No. 668540250MG
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Highly selective organocatalyst for the Diels-Alder reaction with simple +a,+a-unsaturated ketones.U.S. Pat. 6,369,243 and related patents apply. For research purposes only.

Specifications

CAS 415678-40-9
Molecular Formula C16H18N2O2
Refractive Index n20/D 1.5598
Linear Formula C16H18N2O2
MDL Number MFCD08457653
Quantity 250 mg
Synonym (2S,5S)-3-Methyl-2-(5-methyl-2-furanyl)-5-(phenylmethyl)-4-imidazolidinone
Molecular Weight (g/mol) 270.33 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.