Learn More
Medchemexpress LLC (2S,5S)-(E,E)-8-(5-(4-(trifluoromethyl)phenyl)-2,4-pentadienoylamino)benzolactam | 497259-23-1 | MFCD09037558 | 68.0% | 501.54 g/mol | C27H30F3N3O3 | 5MG
SDP
Catalog No. 5000237364
Encompass_Preferred
TPPB is a benzolactam-derived, cell-permeable protein kinase C (PKC) activator used in biochemical and cell-based research. It exhibits high affinity for PKC with a reported Ki of 11.9 nM and is applied in studies of cell differentiation and amyloid precursor protein modulation. This compound is intended for research use only and should be handled according to safety data guidelines.
  • High-affinity PKC activation (Ki = 11.9 nM).
  • Cell-permeable small molecule suitable for cellular assays.
  • Applicable to studies of differentiation and APP modulation.
  • Supplied in small quantities for research applications.
  • Handle according to safety data sheet instructions.
Catalog No. 50-002-37364 Supplier Medchemexpress LLC Supplier No. HY123595MG
TPPB is a benzolactam-derived, cell-permeable protein kinase C (PKC) activator used in biochemical and cell-based research. It exhibits high affinity for PKC with a reported Ki of 11.9 nM and is applied in studies of cell differentiation and amyloid precursor protein modulation. This compound is intended for research use only and should be handled according to safety data guidelines.
  • High-affinity PKC activation (Ki = 11.9 nM).
  • Cell-permeable small molecule suitable for cellular assays.
  • Applicable to studies of differentiation and APP modulation.
  • Supplied in small quantities for research applications.
  • Handle according to safety data sheet instructions.