Learn More
Cayman Chemical 2S-6-PrenylnarngenIn 10mg
SDP
Catalog No. 502912391
Encompass_Preferred
A prenylated flavonoid with diverse biological activities; inhibits the Cav3.2 channel in HEK293 cells and high-voltage-activated calcium currents in differentiated NG 108-15 cells in whole-cell patch-clamp assays (IC50s = 0.928 and 2.005 µM, respectively); reduces the activity of ALR2 (IC50 = 6.2 µM for the recombinant human enzyme); reduces mechanical allodynia in a mouse model of sodium hydrosulfide-induced somatic pain at 10 pmol/paw
Catalog No. 50-291-2391 Supplier Cayman Chemical Supplier No. 3513810 MG
A prenylated flavonoid with diverse biological activities; inhibits the Cav3.2 channel in HEK293 cells and high-voltage-activated calcium currents in differentiated NG 108-15 cells in whole-cell patch-clamp assays (IC50s = 0.928 and 2.005 µM, respectively); reduces the activity of ALR2 (IC50 = 6.2 µM for the recombinant human enzyme); reduces mechanical allodynia in a mouse model of sodium hydrosulfide-induced somatic pain at 10 pmol/paw