Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (2S,6S)-hydroxynorketamine (hydrochloride) | 1430202-70-2 | MFCD30720898 | 99.3% | 276.16 | C12H15Cl2NO2 | 10 MG
SDP

Catalog No. 5000247929
Encompass_Preferred

(2S,6S)-Hydroxynorketamine hydrochloride is the hydrochloride salt of the 2S,6S stereoisomer of hydroxynorketamine, a ketamine metabolite widely used as a research reagent in neuropharmacology and mTOR-related studies. Supplied as a white to off-white solid with high reported purity, it is intended for biochemical and cellular assays and is available in milligram-scale pack sizes for laboratory use.

  • High reported purity: 99.3%.
  • Hydrochloride salt; solid white to off-white appearance.
  • Molecular weight 276.16 g/mol.
  • Suitable for biochemical and cellular assays in neuropharmacology research.
  • Recommended storage: 4°C sealed; in solvent -80°C (6 months) or -20°C (1 month).
  • Available in milligram pack sizes, including 10 mg.

Catalog No. 50-002-47929 Supplier Medchemexpress LLC Supplier No. HY10195510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(2S,6S)-Hydroxynorketamine hydrochloride is the hydrochloride salt of the 2S,6S stereoisomer of hydroxynorketamine, a ketamine metabolite widely used as a research reagent in neuropharmacology and mTOR-related studies. Supplied as a white to off-white solid with high reported purity, it is intended for biochemical and cellular assays and is available in milligram-scale pack sizes for laboratory use.

  • High reported purity: 99.3%.
  • Hydrochloride salt; solid white to off-white appearance.
  • Molecular weight 276.16 g/mol.
  • Suitable for biochemical and cellular assays in neuropharmacology research.
  • Recommended storage: 4°C sealed; in solvent -80°C (6 months) or -20°C (1 month).
  • Available in milligram pack sizes, including 10 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.