Learn More
Medchemexpress LLC (2S,6S)-hydroxynorketamine hydrochloride | 1430202-70-2 | MFCD30720898 | 99.3% | 276.16 g/mol | C12H15Cl2NO2 | 5 MG
SDP
Catalog No. 5000248583
Encompass_Preferred
(2S,6S)-Hydroxynorketamine hydrochloride is the hydrochloride salt of a ketamine metabolite that shows neuroactive effects in preclinical studies, including antidepressant-like and analgesic activities. It modulates intracellular signaling pathways such as mTOR and interacts with nicotinic acetylcholine receptors, making it useful for mechanistic pharmacology research. The compound is supplied as a solid for laboratory use.
  • Ketamine metabolite with neuroactive properties.
  • Modulates mTOR signaling and downstream phosphorylation.
  • Antagonizes α7-nicotinic acetylcholine receptor.
  • Supplied as a white to off-white solid for research use.
  • Available in small-mass pack sizes suitable for preclinical studies.
  • High purity appropriate for analytical and biological assays.
Catalog No. 50-002-48583 Supplier Medchemexpress LLC Supplier No. HY1019555MG
(2S,6S)-Hydroxynorketamine hydrochloride is the hydrochloride salt of a ketamine metabolite that shows neuroactive effects in preclinical studies, including antidepressant-like and analgesic activities. It modulates intracellular signaling pathways such as mTOR and interacts with nicotinic acetylcholine receptors, making it useful for mechanistic pharmacology research. The compound is supplied as a solid for laboratory use.
  • Ketamine metabolite with neuroactive properties.
  • Modulates mTOR signaling and downstream phosphorylation.
  • Antagonizes α7-nicotinic acetylcholine receptor.
  • Supplied as a white to off-white solid for research use.
  • Available in small-mass pack sizes suitable for preclinical studies.
  • High purity appropriate for analytical and biological assays.