Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2S)-Bornane-10,2-sultam 99+%, (99+% e.e.), Thermo Scientific™

Catalog No. AC294180010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC294180010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC294180010 Supplier Thermo Scientific Chemicals Supplier No. 294180010

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108448-77-7
Molecular Formula C10H17NO2S
Molecular Weight (g/mol) 215.31
MDL Number MFCD00151762,MFCD00066271,MFCD00151762,MFCD00151762
InChI Key DPJYJNYYDJOJNO-UHFFFAOYNA-N
Synonym 6s,7as-8,8-dimethylhexahydro-3h-3a,6-methanobenzo c isothiazole 2,2-dioxide
PubChem CID 132274328
IUPAC Name 10,10-dimethyl-3λ⁶-thia-4-azatricyclo[5.2.1.0¹,⁵]decane-3,3-dione
SMILES CC1(C)C2CCC11CS(=O)(=O)NC1C2

Specifications

Melting Point 181°C to 185°C
Color White
Enantiomeric Excess 99.0% min.
Infrared Spectrum Authentic
Quantity 1 g
Specific Rotation +32° to +34° (20°C, 589nm) (c=5, CHCl3)
Formula Weight 215.32
Percent Purity ≥99.0%
Physical Form Flakes
Chemical Name or Material (2S)-Bornane-10, 2-sultam, 99+%, (99+% e.e.)
Hazard Category GHS Signal Word: Warning
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.