Learn More
Cayman Chemical 2S-Eriodictyol, Purity: ≥98%, CAS: 552-58-9, 10 mg
SDP
Catalog No. 502936797
Encompass_Preferred
A flavanone with antioxidant activity; protective against hydrogen peroxide-induced cell injury and apoptosis in EA.hy926 cells at 20 µM
Catalog No. 50-293-6797 Supplier Cayman Chemical Supplier No. 2680310 MG
A flavanone with antioxidant activity; protective against hydrogen peroxide-induced cell injury and apoptosis in EA.hy926 cells at 20 µM