Learn More
Cayman Chemical 2S-ErIodIctyol 25mg
SDP
Catalog No. 502932972
Encompass_Preferred
A flavanone with antioxidant activity; protective against hydrogen peroxide-induced cell injury and apoptosis in EA.hy926 cells at 20 µM
Catalog No. 50-293-2972 Supplier Cayman Chemical Supplier No. 2680325 MG
A flavanone with antioxidant activity; protective against hydrogen peroxide-induced cell injury and apoptosis in EA.hy926 cells at 20 µM