Learn More
Echelon Biosciences Research Labs (2S)-ETHER LPA 1 mg
SDP
Supplier:  Echelon Biosciences Research Labs L01861MG
Encompass
(2S)-Oleyl LPA is an analog of 18:1 LPA in which the fatty ester bond to gycerol has been replaced by a metabolically stable ether linkage. Lysophosphatidic Acid (LPA) is a small lysophospholipid involved in diverse cellular processes such as cell proliferation, chemotaxis, platelet aggregation, wound healing, angiogenesis, tumor invasion, and smooth muscle contraction. LPA binds to several G-coupled protein receptors to initiate its biological functions. In cancer, LPA primarily promotes cell survival, migration and invasion. This item is non returnable
Catalog No. 50-720-480