Learn More
Medchemexpress LLC (2S)-Gemcitabine-13C,15N2 hydrochloride | 1262897-74-4 | 99.6% | C813CH12ClF2N15N2O4 | 1 MG
SDP
Catalog No. 5000432397
Encompass_Preferred
(2S)-Gemcitabine-13C,15N2 hydrochloride is a 13C and 15N labeled version of (2S)-Gemcitabine hydrochloride. It is intended for research use only. Stable heavy isotopes, such as those incorporated into this drug molecule, are used as tracers for quantitation during drug development, with deuteration specifically noted for its potential impact on pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-32397 Supplier Medchemexpress LLC Supplier No. HYB0003AS1MG
(2S)-Gemcitabine-13C,15N2 hydrochloride is a 13C and 15N labeled version of (2S)-Gemcitabine hydrochloride. It is intended for research use only. Stable heavy isotopes, such as those incorporated into this drug molecule, are used as tracers for quantitation during drug development, with deuteration specifically noted for its potential impact on pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.