Learn More
Medchemexpress LLC HY-N2584 5mg Medchemexpress, (2S)-Isoxanthohumol CAS:70872-29-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN25845MG
Encompass_Preferred
Medchemexpress, HY-N2584 5mg (2S)-Isoxanthohumol CAS:70872-29-6 (2S)-Isoxanthohumol is a microbial biotransformed metabolite of the hop prenylflavanone Isoxanthohumol. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2907