Learn More
Medchemexpress LLC (2S)-Isoxanthohumol | 70872-29-6 | 1 MG
SDP
Catalog No. 5000420736
Encompass_Preferred
(2S)-Isoxanthohumol is a microbial biotransformed metabolite of the hop prenylflavanone Isoxanthohumol. It is for research use only and not sold to patients.
  • Exhibits antineuroinflammatory activity in mouse BV2 cells, assessed as inhibition of LPS-induced nitric oxide production.
Catalog No. 50-004-20736 Supplier Medchemexpress LLC Supplier No. HYN25841MG
(2S)-Isoxanthohumol is a microbial biotransformed metabolite of the hop prenylflavanone Isoxanthohumol. It is for research use only and not sold to patients.
  • Exhibits antineuroinflammatory activity in mouse BV2 cells, assessed as inhibition of LPS-induced nitric oxide production.