Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2S)-N-[(1S)-1-(Hydroxydiphenylmethyl)-3-methylbutyl]-2-pyrrolidinecarboxamide 98.0+%, TCI America™
SDP

Catalog No. H1407200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H1407200MG 200 mg
H14071G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H1407200MG Supplier TCI America Supplier No. H1407200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 910110-45-1
Molecular Formula C23H30N2O2
Molecular Weight (g/mol) 366.505
MDL Number MFCD22200506
InChI Key PWBULUUPVDXEQS-SFTDATJTSA-N
Synonym (S)-N-[(S)-1-Hydroxy-4-methyl-1,1-diphenylpentan-2-yl]pyrrolidine-2-carboxamide, N-[(1S)-1-(Hydroxydiphenylmethyl)-3-methylbutyl]-L-prolinamide, Singh′Cs Catalyst
PubChem CID 11954678
IUPAC Name (2S)-N-[(2S)-1-hydroxy-4-methyl-1,1-diphenylpentan-2-yl]pyrrolidine-2-carboxamide
SMILES CC(C)CC(C(C1=CC=CC=C1)(C2=CC=CC=C2)O)NC(=O)C3CCCN3

Specifications

Melting Point 193°C
Color White
Quantity 200 mg
Formula Weight 366.51
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (2S)-N-[(1S)-1-(Hydroxydiphenylmethyl)-3-methylbutyl]-2-pyrrolidinecarboxamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.