Learn More
Medchemexpress LLC (2S)-N-[7-chloro-6-[4-[(3R,4R)-4-hydroxy-3-methyloxolan-3-yl]piperazin-1-yl]isoquinolin-3-yl]-6-o... | 2769107-89-1 | 99.7% | 501.02 g·mol⁻1 | C26H33ClN4O4 | 10 MG
SDP
Catalog No. 5000288091
Encompass_Preferred
MK-1468 is an orally active, selective, blood-brain barrier-permeable inhibitor of leucine-rich repeat kinase 2 (LRRK2) used in preclinical research of Parkinson's disease and LRRK2-related pathways.
  • Orally active and brain-penetrant LRRK2 inhibitor suitable for in vivo and in vitro studies.
  • High purity (≈99.7%) for research-grade experiments.
  • Molecular weight 501.02 g·mol⁻1 and formula C26H33ClN4O4.
  • Available in small-scale quantities for dosing and assay optimization.
  • Store powder at -20°C; solutions stable at -80°C for 6 months.
Catalog No. 50-002-88091 Supplier Medchemexpress LLC Supplier No. HY15642910MG
MK-1468 is an orally active, selective, blood-brain barrier-permeable inhibitor of leucine-rich repeat kinase 2 (LRRK2) used in preclinical research of Parkinson's disease and LRRK2-related pathways.
  • Orally active and brain-penetrant LRRK2 inhibitor suitable for in vivo and in vitro studies.
  • High purity (≈99.7%) for research-grade experiments.
  • Molecular weight 501.02 g·mol⁻1 and formula C26H33ClN4O4.
  • Available in small-scale quantities for dosing and assay optimization.
  • Store powder at -20°C; solutions stable at -80°C for 6 months.