Learn More
Medchemexpress LLC (2S)-N-methyl-2-[[2-(2-methyl-1H-indol-3-yl)acetyl]amino]-N,3-diphenylpropanamide | 1352879-65-2 | 98.9% | 10 MG
SDP
Catalog No. 5000234312
Encompass_Preferred
PF-3450074 (PF-74) is a small-molecule HIV-1 capsid inhibitor supplied as a high-purity powder for research use. It is commonly used as a biochemical probe in virology and antiviral mechanism studies.
  • High purity suitable for biochemical and cellular assays.
  • Research-grade powder form with recommended DMSO solubility.
  • Molecular weight 425.52 g·mol⁻¹ and formula C27H27N3O2.
  • Stable when stored at low temperatures for long-term use.
  • Intended for research use only, not for clinical applications.
Catalog No. 50-002-34312 Supplier Medchemexpress LLC Supplier No. HY12007210MG
PF-3450074 (PF-74) is a small-molecule HIV-1 capsid inhibitor supplied as a high-purity powder for research use. It is commonly used as a biochemical probe in virology and antiviral mechanism studies.
  • High purity suitable for biochemical and cellular assays.
  • Research-grade powder form with recommended DMSO solubility.
  • Molecular weight 425.52 g·mol⁻¹ and formula C27H27N3O2.
  • Stable when stored at low temperatures for long-term use.
  • Intended for research use only, not for clinical applications.