Learn More
Medchemexpress LLC (2S)-N-methyl-2-[[2-(2-methyl-1H-indol-3-yl)acetyl]amino]-N,3-diphenylpropanamide | 1352879-65-2 | 98.9% | 100 MG
SDP
Catalog No. 5000233177
Encompass_Preferred
PF-3450074 (PF-74) is a small-molecule inhibitor of the HIV-1 capsid protein for in vitro research. It shows broad-spectrum inhibition with submicromolar potency and is supplied as a powder and as a 10 mM DMSO solution for laboratory assays.
  • Broad-spectrum HIV-1 capsid inhibition with submicromolar potency.
  • High purity (98.9%) suitable for biochemical assays.
  • Available as powder (multiple sizes) and 10 mM DMSO solution.
  • Molecular formula C27H27N3O2 and molecular weight 425.52 g/mol for database reference.
  • Stable under recommended storage conditions for long-term use.
Catalog No. 50-002-33177 Supplier Medchemexpress LLC Supplier No. HY120072100MG
PF-3450074 (PF-74) is a small-molecule inhibitor of the HIV-1 capsid protein for in vitro research. It shows broad-spectrum inhibition with submicromolar potency and is supplied as a powder and as a 10 mM DMSO solution for laboratory assays.
  • Broad-spectrum HIV-1 capsid inhibition with submicromolar potency.
  • High purity (98.9%) suitable for biochemical assays.
  • Available as powder (multiple sizes) and 10 mM DMSO solution.
  • Molecular formula C27H27N3O2 and molecular weight 425.52 g/mol for database reference.
  • Stable under recommended storage conditions for long-term use.