Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (2S)-N-methyl-2-[[2-(2-methyl-1H-indol-3-yl)acetyl]amino]-N,3-diphenylpropanamide | 1352879-65-2 | 98.9% | 25 MG
SDP

Catalog No. 5000233922
Encompass_Preferred

PF-3450074 (PF-74) is a small-molecule inhibitor of the HIV-1 capsid protein used in laboratory research; it shows broad-spectrum, submicromolar inhibition and interferes with capsid-host factor interactions to block uncoating, assembly, and reverse transcription.

  • High purity suitable for research use (98.9%).
  • Molecular formula C27H27N3O2; molecular weight 425.52 g·mol-1.
  • Soluble in DMSO (up to 250 mg/mL); example in vivo formulation: 10% DMSO, 90% corn oil.
  • Recommended storage: powder at -20°C for long term, 4°C for short term; stock solutions at -80°C.
  • Intended for laboratory research only, not for human use.

Catalog No. 50-002-33922 Supplier Medchemexpress LLC Supplier No. HY12007225MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PF-3450074 (PF-74) is a small-molecule inhibitor of the HIV-1 capsid protein used in laboratory research; it shows broad-spectrum, submicromolar inhibition and interferes with capsid-host factor interactions to block uncoating, assembly, and reverse transcription.

  • High purity suitable for research use (98.9%).
  • Molecular formula C27H27N3O2; molecular weight 425.52 g·mol-1.
  • Soluble in DMSO (up to 250 mg/mL); example in vivo formulation: 10% DMSO, 90% corn oil.
  • Recommended storage: powder at -20°C for long term, 4°C for short term; stock solutions at -80°C.
  • Intended for laboratory research only, not for human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.