Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (2S)-N-methyl-2-[[2-(2-methyl-1H-indol-3-yl)acetyl]amino]-N,3-diphenylpropanamide | 1352879-65-2 | 98.9% | 50 MG
SDP

Catalog No. 5000233568
Encompass_Preferred

PF-3450074 (PF-74) is a small-molecule inhibitor of the HIV-1 capsid protein that disrupts capsid function and inhibits early stages of viral replication, making it useful for mechanistic and antiviral research.

  • Specific inhibitor of HIV-1 capsid protein, acting at early stages of infection.
  • IUPAC name: (2S)-N-methyl-2-[[2-(2-methyl-1H-indol-3-yl)acetyl]amino]-N,3-diphenylpropanamide.
  • Molecular formula C27H27N3O2; molecular weight 425.52 g/mol; purity 98.9%.
  • Soluble in DMSO (250 mg/mL); in vivo vehicle example: 10% DMSO, 90% corn oil (≥ 6.25 mg/mL).
  • Recommended storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (2 years) or -20°C (1 year).
  • Supplied in small research quantities; this item is provided as a 50 mg package.

Catalog No. 50-002-33568 Supplier Medchemexpress LLC Supplier No. HY12007250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PF-3450074 (PF-74) is a small-molecule inhibitor of the HIV-1 capsid protein that disrupts capsid function and inhibits early stages of viral replication, making it useful for mechanistic and antiviral research.

  • Specific inhibitor of HIV-1 capsid protein, acting at early stages of infection.
  • IUPAC name: (2S)-N-methyl-2-[[2-(2-methyl-1H-indol-3-yl)acetyl]amino]-N,3-diphenylpropanamide.
  • Molecular formula C27H27N3O2; molecular weight 425.52 g/mol; purity 98.9%.
  • Soluble in DMSO (250 mg/mL); in vivo vehicle example: 10% DMSO, 90% corn oil (≥ 6.25 mg/mL).
  • Recommended storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (2 years) or -20°C (1 year).
  • Supplied in small research quantities; this item is provided as a 50 mg package.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.