Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (2S)-N,N-dimethyl-5-(1,3,5-trimethylpyrazol-4-yl)-1,2,3,4-tetrahydronaphthalen-2-amine | 1000578-26-6 | MFCD09878234 | 99.7% | 283.41 | C18H25N3 | 50 MG
SDP

Catalog No. 5000245924
Encompass_Preferred

Potent, selective 5-HT7 receptor agonist used in neuroscience research to study memory consolidation and serotonergic signaling. It has subnanomolar binding affinity and demonstrated activity in vitro and in vivo models.

  • Potent 5-HT7 receptor agonist with IC50 ≈ 0.83 nM and Ki ≈ 0.6 nM.
  • Selective versus other serotonin receptor subtypes at relevant concentrations.
  • High reported purity (~99.7%).
  • Appearance: oil, light yellow to brown.
  • Supplied in 50 MG research quantities for laboratory use.
  • Recommended storage: -20°C (dry) for long-term stability; -80°C in solution for extended storage.

Catalog No. 50-002-45924 Supplier Medchemexpress LLC Supplier No. HY10314250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent, selective 5-HT7 receptor agonist used in neuroscience research to study memory consolidation and serotonergic signaling. It has subnanomolar binding affinity and demonstrated activity in vitro and in vivo models.

  • Potent 5-HT7 receptor agonist with IC50 ≈ 0.83 nM and Ki ≈ 0.6 nM.
  • Selective versus other serotonin receptor subtypes at relevant concentrations.
  • High reported purity (~99.7%).
  • Appearance: oil, light yellow to brown.
  • Supplied in 50 MG research quantities for laboratory use.
  • Recommended storage: -20°C (dry) for long-term stability; -80°C in solution for extended storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.