Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (2S)-N,N-dimethyl-5-(1,3,5-trimethylpyrazol-4-yl)-1,2,3,4-tetrahydronaphthalen-2-amine | 1000578-26-6 | MFCD09878234 | 99.7% | 283.41 g·mol⁻1 | C18H25N3 | 10 MG
SDP

Catalog No. 5000246231
Encompass_Preferred

AS19 is a potent, selective 5-HT7 receptor agonist used in neuroscience research to study memory consolidation and serotonergic signaling. It offers subnanomolar potency and is supplied at high purity for research applications.

  • Potent and selective 5-HT7 receptor agonist (human IC50 0.83 nM).
  • High purity (~99.7%).
  • Molecular weight 283.41 g·mol⁻1; formula C18H25N3.
  • Available in research pack sizes from 1 mg to 100 mg and as 10 mM in DMSO.
  • Recommended storage: pure form -20°C (stable ~3 years); in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-46231 Supplier Medchemexpress LLC Supplier No. HY10314210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AS19 is a potent, selective 5-HT7 receptor agonist used in neuroscience research to study memory consolidation and serotonergic signaling. It offers subnanomolar potency and is supplied at high purity for research applications.

  • Potent and selective 5-HT7 receptor agonist (human IC50 0.83 nM).
  • High purity (~99.7%).
  • Molecular weight 283.41 g·mol⁻1; formula C18H25N3.
  • Available in research pack sizes from 1 mg to 100 mg and as 10 mM in DMSO.
  • Recommended storage: pure form -20°C (stable ~3 years); in solvent -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.