Learn More
Medchemexpress LLC (2S)-Octyl-α-hydroxyglutarate | 1391194-64-1 | 95.0% | 260.33 g/mol | C13H24O5 | 1 ML
SDP
Catalog No. 5000244587
Encompass_Preferred
(2S)-Octyl-α-hydroxyglutarate is a cell-permeable octyl-ester derivative of the S-isomer of 2-hydroxyglutarate supplied for research use. It is commonly provided as a 10 mM solution in DMSO (1 mL vial) and is used as a cell-permeant precursor to L-2-hydroxyglutarate in cellular and epigenetic studies.
  • Cell-permeable octyl ester of S-2-hydroxyglutarate.
  • Provided as a 10 mM solution in DMSO, 1 mL vial.
  • Molecular formula C13H24O5; molecular weight 260.33 g/mol.
  • Intended for in vitro research use only; not for human or clinical use.
  • Used in studies of cellular metabolism and histone methylation.
Catalog No. 50-002-44587 Supplier Medchemexpress LLC Supplier No. HY103641A10MM 1ML
(2S)-Octyl-α-hydroxyglutarate is a cell-permeable octyl-ester derivative of the S-isomer of 2-hydroxyglutarate supplied for research use. It is commonly provided as a 10 mM solution in DMSO (1 mL vial) and is used as a cell-permeant precursor to L-2-hydroxyglutarate in cellular and epigenetic studies.
  • Cell-permeable octyl ester of S-2-hydroxyglutarate.
  • Provided as a 10 mM solution in DMSO, 1 mL vial.
  • Molecular formula C13H24O5; molecular weight 260.33 g/mol.
  • Intended for in vitro research use only; not for human or clinical use.
  • Used in studies of cellular metabolism and histone methylation.