Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (2S)-tert-butoxy[4-(2,3-dihydropyrano[4,3,2-de]quinolin-7-yl)-2-methylquinolin-3-yl]acetic acid | 1155419-89-8 | 99.7% | 442.51 g/mol | C27H26N2O4 | 10 MG
SDP

Catalog No. 5000295050
Encompass_Preferred

BI 224436 is a research-grade small-molecule noncatalytic site integrase inhibitor for HIV-1, intended for in vitro antiviral and biochemical studies. It demonstrates potent activity against laboratory strains and is supplied as a solid with defined storage requirements.

  • Potent noncatalytic site integrase inhibitor with EC50 <15 nM.
  • High purity, 99.7%.
  • Molecular weight 442.51 g/mol.
  • Chemical formula C27H26N2O4.
  • Solid, white to yellow appearance.
  • Store at -20°C and protect from light.
  • Suitable for in vitro antiviral and biochemical assays.

Catalog No. 50-002-95050 Supplier Medchemexpress LLC Supplier No. HY1859510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BI 224436 is a research-grade small-molecule noncatalytic site integrase inhibitor for HIV-1, intended for in vitro antiviral and biochemical studies. It demonstrates potent activity against laboratory strains and is supplied as a solid with defined storage requirements.

  • Potent noncatalytic site integrase inhibitor with EC50 <15 nM.
  • High purity, 99.7%.
  • Molecular weight 442.51 g/mol.
  • Chemical formula C27H26N2O4.
  • Solid, white to yellow appearance.
  • Store at -20°C and protect from light.
  • Suitable for in vitro antiviral and biochemical assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.