Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical 1-(Fmoc-aminomethyl)-beta-D-galacturonic acid | 100 g | CAS 655246-28-9 | MDL MFCD11974983

Catalog No. 502343178
Encompass

1-(Fmoc-aminomethyl)-beta-D-galacturonic acid is a Amino Acid reagent (Subcategory: Sugar AA) sold by WuXi TIDES. Offered in 100 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 655246-28-9
- MDL: MFCD11974983
- InChIKey: USLKEBRNVKYNDJ-OMZCGLGVSA-N
- Molecular Weight: 429.425
- Molecular Formula: C22H23NO8
- Purity: ≥95%
- Container Type: 500 mL HDPE
- Pack Size: 100 g
- Net Weight: 100 g
- Gross Weight: 160.5 g
- Commodity Code: 29329900
- Country Of Origin: China
- IUPAC: (2S,3R,4R,5R,6S)-6-(((((9H-fluoren-9-yl)methoxy)carbonyl)amino)methyl)-3,4,5-trihydroxytetrahydro-2H-pyran-2-carboxylic acid
- SMILES: O=C([C@H]1O[C@@H](CNC(OCC2C3=CC=CC=C3C4=CC=CC=C42)=O)[C@H](O)[C@@H](O)[C@H]1O)O

Catalog No. 50-234-3178 Supplier STA Pharmaceutical Supplier No. STAAA12022100GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-(Fmoc-aminomethyl)-beta-D-galacturonic acid is a Amino Acid reagent (Subcategory: Sugar AA) sold by WuXi TIDES. Offered in 100 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 655246-28-9
- MDL: MFCD11974983
- InChIKey: USLKEBRNVKYNDJ-OMZCGLGVSA-N
- Molecular Weight: 429.425
- Molecular Formula: C22H23NO8
- Purity: ≥95%
- Container Type: 500 mL HDPE
- Pack Size: 100 g
- Net Weight: 100 g
- Gross Weight: 160.5 g
- Commodity Code: 29329900
- Country Of Origin: China
- IUPAC: (2S,3R,4R,5R,6S)-6-(((((9H-fluoren-9-yl)methoxy)carbonyl)amino)methyl)-3,4,5-trihydroxytetrahydro-2H-pyran-2-carboxylic acid
- SMILES: O=C([C@H]1O[C@@H](CNC(OCC2C3=CC=CC=C3C4=CC=CC=C42)=O)[C@H](O)[C@@H](O)[C@H]1O)O

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.