Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical (2S,3S)-1-(((9H-fluoren-9-yl)methoxy)carbonyl)-3-phenylpyrrolidine-2-carboxylic acid (Abs) | 25 g | CAS 1158891-09-8 | InChIKey XZINBBVSLWSWBO-KOSHJBKYSA-N

Catalog No. 502353040
Encompass

(2S,3S)-1-(((9H-fluoren-9-yl)methoxy)carbonyl)-3-phenylpyrrolidine-2-carboxylic acid (Abs) is a Amino Acid reagent (Subcategory: Pro) sold by WuXi TIDES. Offered in 25 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 1158891-09-8
- MDL: No data
- InChIKey: XZINBBVSLWSWBO-KOSHJBKYSA-N
- Molecular Weight: 413.473
- Molecular Formula: C26H23NO4
- Purity: ≥95%
- Container Type: 125 mL HDPE
- Pack Size: 25 g
- Net Weight: 25 g
- Gross Weight: 54.3 g
- Commodity Code: 29339980
- Country Of Origin: China
- IUPAC: (2S,3R)-1-(((9H-fluoren-9-yl)methoxy)carbonyl)-3-phenylpyrrolidine-2-carboxylic acid
- SMILES: OC(=O)[C@@H]1[C@H](CCN1C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C1=CC=CC=C1

Catalog No. 50-235-3040 Supplier STA Pharmaceutical Supplier No. STAAA1016525GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(2S,3S)-1-(((9H-fluoren-9-yl)methoxy)carbonyl)-3-phenylpyrrolidine-2-carboxylic acid (Abs) is a Amino Acid reagent (Subcategory: Pro) sold by WuXi TIDES. Offered in 25 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 1158891-09-8
- MDL: No data
- InChIKey: XZINBBVSLWSWBO-KOSHJBKYSA-N
- Molecular Weight: 413.473
- Molecular Formula: C26H23NO4
- Purity: ≥95%
- Container Type: 125 mL HDPE
- Pack Size: 25 g
- Net Weight: 25 g
- Gross Weight: 54.3 g
- Commodity Code: 29339980
- Country Of Origin: China
- IUPAC: (2S,3R)-1-(((9H-fluoren-9-yl)methoxy)carbonyl)-3-phenylpyrrolidine-2-carboxylic acid
- SMILES: OC(=O)[C@@H]1[C@H](CCN1C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C1=CC=CC=C1

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.