Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical (2S,4S)-1-(((9H-fluoren-9-yl)methoxy)carbonyl)-4-(((allyloxy)carbonyl)amino)pyrrolidine-2-carboxylic acid | 1 g | CAS 274676-10-7 | MDL MFCD22989434

Catalog No. 502352413
Encompass

(2S,4S)-1-(((9H-fluoren-9-yl)methoxy)carbonyl)-4-(((allyloxy)carbonyl)amino)pyrrolidine-2-carboxylic acid is a Amino Acid reagent (Subcategory: Pro) sold by WuXi TIDES. Offered in 1 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 274676-10-7
- MDL: MFCD22989434
- InChIKey: DSGCPQRUOSTFEF-BTYIYWSLSA-N
- Molecular Weight: 436.464
- Molecular Formula: C24H24N2O6
- Purity: ≥95%
- Container Type: 15 mL HDPE
- Pack Size: 1 g
- Net Weight: 1 g
- Gross Weight: 7.8 g
- Commodity Code: 29339980
- Country Of Origin: China
- IUPAC: (2S,4S)-1-(((9H-fluoren-9-yl)methoxy)carbonyl)-4-(((allyloxy)carbonyl)amino)pyrrolidine-2-carboxylic acid
- SMILES: C=CCOC(N[C@H]1C[C@H](N(C1)C(OCC2C3=CC=CC=C3C4=C2C=CC=C4)=O)C(O)=O)=O

Catalog No. 50-235-2413 Supplier STA Pharmaceutical Supplier No. STAAA101741GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(2S,4S)-1-(((9H-fluoren-9-yl)methoxy)carbonyl)-4-(((allyloxy)carbonyl)amino)pyrrolidine-2-carboxylic acid is a Amino Acid reagent (Subcategory: Pro) sold by WuXi TIDES. Offered in 1 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 274676-10-7
- MDL: MFCD22989434
- InChIKey: DSGCPQRUOSTFEF-BTYIYWSLSA-N
- Molecular Weight: 436.464
- Molecular Formula: C24H24N2O6
- Purity: ≥95%
- Container Type: 15 mL HDPE
- Pack Size: 1 g
- Net Weight: 1 g
- Gross Weight: 7.8 g
- Commodity Code: 29339980
- Country Of Origin: China
- IUPAC: (2S,4S)-1-(((9H-fluoren-9-yl)methoxy)carbonyl)-4-(((allyloxy)carbonyl)amino)pyrrolidine-2-carboxylic acid
- SMILES: C=CCOC(N[C@H]1C[C@H](N(C1)C(OCC2C3=CC=CC=C3C4=C2C=CC=C4)=O)C(O)=O)=O

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.