Learn More
Cayman Chemical (2S)-Octyl-α-hydroxyglutarate, Purity: ≥95%, CAS: 1391194-64-1, 10 mg
SDP
Catalog No. 502968810
Encompass_Preferred
A cell-permeable derivative of the L-isomer of 2-HG
Catalog No. 50-296-8810 Supplier Cayman Chemical Supplier No. 1636710 MG
A cell-permeable derivative of the L-isomer of 2-HG