Learn More
CPC Scientific 3-Mercaptopropionyl-D-Tyr(Et)-Ile-Thr-Asn-Cys-Pro-Orn-Gly-NH2 (acetate salt) 250MG
SDP
Supplier:  CPC Scientific OXYT003B
Encompass
vasopressin receptor antagonist.ONE-LETTER SEQUENCE: 3-Mercaptopropionyl-y(Et)-ITNCP-Orn-G-NH2MOLECULAR FORMULA: C43H67N11O12S2MOLECULAR WEIGHT:994.2STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [90779-69-4], RESEARCH AREA: HormonalREFERENCES: M.Akerlund, Prog. Brain Res., 139, 359 (2002); H.H.Zingg and S.A.Laporte, Trends Endocrinol. Metab., 14, 222 (2003); M.Manning et al., Prog. Brain Res., 170, 473 (2008)
Catalog No. 50-211-0049